Skip to Content
Merck
All Photos(1)

Key Documents

P0025

Sigma-Aldrich

Platensimycin

≥90% (HPLC), from Streptomyces platensis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO7
CAS Number:
Molecular Weight:
441.47
UNSPSC Code:
12161606
PubChem Substance ID:

biological source

Streptomyces platensis

Assay

≥90% (HPLC)

form

solid

color

white to beige

solubility

DMSO: 1 mg/mL, clear, colorless

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

O=C1C=C[C@]2(C[H])[C@H]3CC[C@@](O3)([H])C2C1CCC(CNC4=C(O)C=CC(C(O)=O)=C4O)=O

InChI

1S/C22H25NO7/c1-22-9-8-14(25)12(18(22)16-6-7-17(22)30-16)3-2-11(24)10-23-19-15(26)5-4-13(20(19)27)21(28)29/h4-5,8-9,12,16-18,23,26-27H,2-3,6-7,10H2,1H3,(H,28,29)/t12?,16-,17+,18?,22+/m0/s1

InChI key

IGBNLKVUOHQLTO-MBQNLBDGSA-N

Application

Platensimycin (PTM) is a broad-spectrum antibiotic produced by LStreptomyces platensis. Platensimycin is used to inhibit lipid biosynthesis. It is used in diabetes research and to study the potential treatment of metabolic disorders.

Biochem/physiol Actions

Demonstrates no cross-resistance to other antibiotic-resistant bacteria, including MRSA (methicillin-resistant Staphylococcus aureus).
Mode of action: Selectively inhibits lipid biosynthesis by targeting FabF/B within the fatty acid synthesis pathway.
Antimicrobial spectrum: Gram-positive bacteria.
Platensimycin selectively inhibits the elongation-condensing enzymes FabF/B of the fatty acid biosynthesis pathway in bacteria. PTM is a potent and highly selective inhibitor of mammalian fatty acid synthase. Platensimycin specifically inhibits fatty acid synthesis but not sterol synthesis in rat primary hepatocytes. It is thought that the thiol group of FabF Cys163 may be activated through the dipole moment of helix N-α-3 which lowers the pKa. It does not demonstrate cross-resistance to other antibiotic-resistant bacteria, including MRSA (methicillin-resistant Staphylococcus aureus). It is active against Gram-positive bacteria.
Selectively inhibits lipid biosynthesis by targeting FabF/B within the fatty acid synthesis pathway.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Szu-Yi Hsu et al.
ACS synthetic biology, 8(10), 2397-2403 (2019-09-06)
Serofendic acid is a natural neuroprotective molecule found in fetal calf serum. It is able to protect neurons against mechanisms of cell death associated with neurodegenerative disease. Because only trace quantities are present in fetal calf serum and complete chemical
Allen C Price et al.
Journal of bacteriology, 185(14), 4136-4143 (2003-07-03)
The beta-ketoacyl-acyl carrier protein synthases are members of the thiolase superfamily and are key regulators of bacterial fatty acid synthesis. As essential components of the bacterial lipid metabolic pathway, they are an attractive target for antibacterial drug discovery. We have
Makoto Hibi et al.
Communications biology, 4(1), 16-16 (2021-01-06)
The high-valent iron-oxo species formed in the non-heme diiron enzymes have high oxidative reactivity and catalyze difficult chemical reactions. Although the hydroxylation of inert methyl groups is an industrially promising reaction, utilizing non-heme diiron enzymes as such a biocatalyst has
Ian Rowe et al.
International journal of molecular sciences, 16(8), 17909-17932 (2015-08-08)
Membrane permeability is a desired property in drug design, but there have been difficulties in quantifying the direct drug partitioning into native membranes. Platensimycin (PL) is a new promising antibiotic whose biosynthetic production is costly. Six dialkylamine analogs of PL
Kenzie A Clark et al.
Journal of the American Chemical Society, 141(27), 10610-10615 (2019-06-28)
The biosynthetic pathways of microbial natural products provide a rich source of novel enzyme-catalyzed transformations. Using a new bioinformatic search strategy, we recently identified an abundance of gene clusters for ribosomally synthesized and post-translationally modified peptides (RiPPs) that contain at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service