O8751
Oxalic acid diammonium salt hydrate
Synonym(s):
Ammonium oxalate, Ethanedioic acid diammonium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H2O4 · 2NH3 · H2O
CAS Number:
Molecular Weight:
142.11
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
SMILES string
N.OC(=O)C(O)=O
InChI
1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2
InChI key
MSMNVXKYCPHLLN-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y Fan et al.
Genetika, 49(4), 439-447 (2013-07-23)
Rapeseed (Brassica napus L.) is one of the most important economic crops worldwide, and Sclerotinia sclerotiorum is the most dangerous disease that affects its yield greatly. Lipid transfer protein (LTP) has broad-spectrum anti-bacterial and fungal activities. In this study, B.
F Debela et al.
Journal of environmental management, 116, 156-162 (2013-01-15)
Pyromorphite (PY) and some zinc phosphates (Zn-P) are very sparingly soluble minerals and hence can immobilize Pb and Zn in contaminated soils. However, mechanisms leading to the poor efficiency of PY and Zn-P formation in contaminated soils amended with P
Magnetic labeling of proteins for atomic force microscopy.
V V Egorov et al.
Doklady. Biochemistry and biophysics, 448, 33-35 (2013-03-13)
Hong-Joo Lee et al.
Bioresource technology, 130, 211-217 (2013-01-12)
The present study investigated the feasibility of the recovery and reuse oxalic acid in a multistage process for the pretreatment of a lignocellulosic biomass. Electrodialysis (ED), an electrochemical process using ion exchange membranes, was used to recover and reuse oxalic
Lu Gao et al.
International journal of phytoremediation, 15(4), 307-319 (2013-03-16)
Phytolacca americana L. can accumulate large amounts of heavy metals in its aerial tissues, especially cadmium (Cd) and manganese (Mn). It has great potential for use in phytoextraction of metals from multi-metal-contaminated soils. This study was conducted to further investigate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service