Skip to Content
Merck
All Photos(1)

Key Documents

O8137

Sigma-Aldrich

Osteoprotegerin/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

OCIF, OPG

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$588.00

$588.00


Check Cart for Availability


Select a Size

Change View
100 μG
$588.00

About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

$588.00


Check Cart for Availability

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

25-50 ng/mL ED50

mol wt

100 kDa by SDS-PAGE (reducing)
calculated mol wt 70.9 kDa

packaging

pkg of 100 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

SMILES string

CCCCCCCCCCC#CCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-10,13-17H2,1H3,(H,19,20)

InChI key

XXUPLYBCNPLTIW-UHFFFAOYSA-N

Gene Information

General description

Osteoprotegerin (OPG) is a member of the TNFR (tumor necrosis factor receptor) superfamily that can act as a decoy receptor for RANKL (receptor activator of nuclear factor κ-B ligand) and TRAIL (tumor necrosis factor-related apoptosis-inducing ligand).[1][2] The primary structure of OPG protein consists of seven distinct domains, four of which corresponds to the extracellular cysteine-rich domains of TNFR proteins and constitutes the soluble OPG. OPG is expressed in a wide variety of tissues including adult heart, lung, kidney and liver. It is secreted both as a monomeric and a dimeric protein.[3]

Biochem/physiol Actions

Binding of soluble osteoprotegerin (OPG) to sRANKL (soluble receptor activator of nuclear factor κ-B ligand) inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage.[1] Changes in the level of serum OPG are associated with atherosclerosis and cardiovascular disorders.[2] It might also be associated with malignancy and plays a regulatory role in multiple myeloma, prostate, breast, bladder and gastric carcinoma.[4] OPG plays a vital role in bone metabolism and is involved in inhibition of ovariectomy-associated bone loss in rats.[3][4]
OPG is a soluble secreted member of the tumor necrosis factor receptor superfamily that lacks any apparent cell-association motifs. It regulates the formation of osteoclasts and is involved in the regulation of bone resorption and remodeling. Human and mouse OPG share approximately 84% and 94% amino acid sequence identity, respectively, with rat OPG.
Soluble secreted member of the tumor necrosis factor receptor superfamily that lacks any apparent cell-association motifs; regulates the formation of osteoclasts and is involved in the regulation of bone resorption and remodeling.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to neutralize apoptosis of mouse L-929 cells treated with 50 ng/ml recombinant human TRAIL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osteoprotegerin secreted by inflammatory and invasive breast cancer cells induces aneuploidy, cell proliferation and angiogenesis.
Goswami S and Sharma-Walia N
BMC Cancer, 15:935 (2015)
Osteoprotegerin: a novel secreted protein involved in the regulation of bone density.
Simonet WS
Cell, 89(2), 309-319 (1997)
Functions of RANKL/RANK/OPG in bone modeling and remodeling.
Boyce BF and Xing L.
Archives of Biochemistry and Biophysics, 473(2), 139-146 (2008)
Polymorphisms in three genes are associated with hemorrhagic stroke.
Liu W
Brain and Behavior, 5(11), e00395-e00395 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service