Skip to Content
Merck
All Photos(1)

Key Documents

O5636

Sigma-Aldrich

9,12-Octadecadiynoic acid

≥98%

Synonym(s):

ODYA, Ro 3-1314

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$233.00

$233.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
1 MG
$233.00

About This Item

Empirical Formula (Hill Notation):
C18H28O2
CAS Number:
Molecular Weight:
276.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

$233.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥98%

form

crystalline

mp

45-46 °C

solubility

water: slightly soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCC#CCC#CCCCCCCCC(O)=O

InChI

1S/C18H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-5,8,11-17H2,1H3,(H,19,20)

InChI key

KDYILQLPKVZDGB-UHFFFAOYSA-N

Biochem/physiol Actions

Potent cyclooxygenase and lipoxygenase inhibitor.

Caution

Oxygen and light sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hitchcock et al.
British journal of pharmacology, 72(4), 689-695 (1981-04-01)
1. 5,8,11,14-Eicosatetraynoic acid (ETYA) inhibited the antigen-induced contractions of tracheal spirals obtained from actively sensitized guinea-pigs. Consistent data were obtained only when the spirals were treated with indomethacin. 2. ETYA did not affect histamine-induced contractions of indomethacin-treated tracheal spirals. 3.
Ning Wang et al.
Frontiers in physiology, 9, 583-583 (2018-06-08)
Background: Perivascular adipose tissue (PVAT) exerts anti-contractile effects on visceral arteries by release of various perivascular relaxing factors (PVRFs) and opening voltage-gated K+ (Kv) channels in vascular smooth muscle cells (VSMCs). Palmitic acid methyl ester (PAME) has been proposed as
David R Colquhoun et al.
Proteomics, 15(12), 2066-2077 (2015-04-29)
Protein acylation plays a critical role in protein localization and function. Acylation is essential for human immunodeficiency virus 1 (HIV-1) assembly and budding of HIV-1 from the plasma membrane in lipid raft microdomains and is mediated by myristoylation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service