Skip to Content
Merck
All Photos(1)

Documents

O4004

Sigma-Aldrich

Octacosanoic acid

~99% (GC)

Synonym(s):

Montanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)26CO2H
CAS Number:
Molecular Weight:
424.74
Beilstein:
1801616
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

~99% (GC)

mp

91-93 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C28H56O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28(29)30/h2-27H2,1H3,(H,29,30)

InChI key

UTOPWMOLSKOLTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Rodríguez et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(1), 89-93 (2002-11-28)
D-003 is a mixture of long-chain fatty acids isolated and purified from sugar cane wax, the major component of which is 1-octacosanoic acid and which possesses effective antiplatelet, antithrombotic and cholesterol-lowering effects. D-003 was suspended in 1% acacia gum solution
R Menéndez et al.
Pharmacological research, 44(4), 299-304 (2001-10-11)
The present study was undertaken to investigate the effects of D003, a mixture of very long chain saturated fatty acids isolated and purified from sugar cane wax, on cholesterol biosynthesis in cultured fibroblasts. Cholesterol biosynthesis is regulated through feedback regulation
Effect of D-003 on a subconvulsive dose of kainic Acid in rats.
Daisy Carbajal et al.
Drugs in R&D, 5(6), 331-336 (2004-11-26)
Zheng-Ming Tao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 32(24), 2604-2606 (2008-03-15)
To investigate the chemical constituents from aerial part of Curcuma wenyujin. Compounds were isolated by repeated column chromatography on silica gel. Their structures were elucidated on the basis of spectral analysis and comparison with literature data. Six compounds were isolated
[Simple quantitative method of direct transesterification of higher fatty acid in biological samples].
V Z Lankin et al.
Voprosy meditsinskoi khimii, 17(3), 331-335 (1971-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service