Skip to Content
Merck
All Photos(1)

Key Documents

O2139

Sigma-Aldrich

N-Oleoyldopamine

>98%, powder

Synonym(s):

OLDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H43NO3
CAS Number:
Molecular Weight:
417.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98%

form

powder

color

white

solubility

DMSO: soluble ≥20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)NCCc1ccc(O)c(O)c1

Biochem/physiol Actions

N-Oleoyldopamine is an endogenous vanilloid; weak CB1 receptor ligand.

Caution

Desiccate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safa Y Almaghrabi et al.
Thrombosis research, 134(2), 412-417 (2014-06-24)
Plant-derived and endogenous vanilloid-like agents exert their effects on cells through transient receptor potential vanilloid-1 (TRPV1). Little is known about the effects of these agents on platelet aggregation. We investigated the effect of various vanilloid-like agents on in-vitro platelet aggregation
Dajeong Ji et al.
Analytical and bioanalytical chemistry, 406(18), 4491-4499 (2014-05-13)
The transient receptor potential vanilloid 1 (TRPV1) channel, a nonselective Ca(2+) and Na(+) channel, is a molecular transducer of nociceptive stimuli. N-Arachidonoyl dopamine (NADA) and N-oleoyldopamine (OLDA), two unsaturated N-acyldopamines, are major activating endogenous TRPV1 ligands and their presence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service