Skip to Content
Merck
All Photos(3)

Key Documents

O0630

Sigma-Aldrich

Octyl α-D-glucopyranoside

≥98% (GC)

Synonym(s):

n-Octyl α-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Beilstein:
4232620
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥98% (GC)

mol wt

292.37 g/mol

storage temp.

−20°C

SMILES string

CCCCCCCCO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14+/m1/s1

InChI key

HEGSGKPQLMEBJL-RGDJUOJXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octyl α-D-glucopyranoside has been used in a study to assess mesogenic structures with interdigitizing alkyl chains. [1] It has also been used in a study to investigate a recirculation procedure involving water removal by product crystallization. [2]

Biochem/physiol Actions

Dialyzable nonionic detergent, suitable for the solubilization and purification of membrane proteins. Can be used for the crystallization of membrane proteins.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Efficient Preparation of Octyl α-D-Glucopyranoside Monohydrate: A Recirculation Procedure Involving Water Removal by Product Crystallisation
Straathof, A., et al.
Starch/Staerke, 40, 229-234 (1988)
Jung-Deog Lee et al.
Organic letters, 7(6), 963-966 (2005-03-12)
[structure: see text] Porphyrin-based molecularly imprinted polymers (MIPs) were prepared for carbohydrate recognition. A urea-appended porphyrin functional monomer was utilized to provide complementary functionality and quality binding sites throughout the polymer. Each porphyrin-based polymer demonstrates high affinity and differential selectivity
G A Jeffrey et al.
Carbohydrate research, 169, 1-11 (1987-11-15)
The crystal structure of octyl alpha-D-glucopyranoside monohydrate, C14H28O6.H2O, is monoclinic, C2, with Z = 4, a = 17.896(2), b = 5.154(1), c = 18.303(2) A, beta = 90.30(1) degrees. The hemihydrate, C14H28O6.0.5 H2O, is also monoclinic, C2, with Z =
Brown, G.M., et al.
Canadian Journal of Chemistry, 48, 2525-2525 (1970)
Douglas L Miller et al.
The Journal of the Acoustical Society of America, 130(5), 3482-3488 (2011-11-18)
Octyl β-D-glucopyranoside (OGP) has been reported to completely inhibit cavitation-induced cell lysis in vitro, possibly by quenching critical free-radical effects. In this study, the influence of OGP on cell lysis in a 60 rpm rotating-tube exposure apparatus was assessed. HL-60

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service