Skip to Content
Merck
All Photos(1)

Key Documents

NCSTUD002

Sigma-Aldrich

MISSION® Synthetic microRNA Inhibitor

cel-miR-243-3p, Negative Control 2, Sequence from Caenorhabditis elegans with no homology to human and mouse gene sequences

Synonym(s):

Synthetic Tough Decoy, sTuD

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$369.00

$369.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 PKG
$369.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$369.00


0 left in stock

Request a Bulk Order

product line

MISSION®

form

solid

mature sequence

CGGUACGAUCGCGGCGGGAUAUC

Sanger mature/minor accession no.

Sanger microRNA accession no.

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Individual synthetic microRNA inhibitors were designed using a proprietary algorithm, which is based on the work of Haraguchi, T, et al. and in collaboration with Dr. Hideo Iba, University of Tokyo.† This algorithm utilizes the tough decoy (TuD) design. miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation.

The MISSION synthetic miRNA Inhibitors are small, double-stranded RNA molecules designed to inhibit a specific mature miRNA. The miRNA inhibitors were designed using the mature miRNA sequence information from miRBase and are 2′-O-methylated RNA duplexes with a miRNA binding site on each strand. Optimal miRNA inhibition is provided after transfection due to the robust secondary structure of the inhibitor.

  • Long lasting inhibition at very low dosage
  • Excellent resistance to cellular nucleases
  • Custom synthesis available for a variety of species

Other Notes

Based on miRBase V19

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service