Skip to Content
Merck
All Photos(1)

Key Documents

N9891

Sigma-Aldrich

Monoclonal Anti-Nebulin antibody produced in mouse

clone NB2, ascites fluid

Synonym(s):

Anti-AMC6, Anti-NEB177D, Anti-NEM2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

NB2, monoclonal

mol wt

antigen 500 kDa

contains

15 mM sodium azide

species reactivity

vertebrates, chicken

technique(s)

indirect immunofluorescence: 1:400 using frozen sections of animal skeletal muscle
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... NEB(101748402)

Related Categories

General description

Nebulin is a 700kDa protein which is expressed in skeletal muscles. It possesses 35 amino acid repeats and a C terminal serine-rich and SH3 domains.
Nebulin is also localized to cardiac muscle. Monoclonal Anti-Nebulin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

Localizes nebulin in skeletal muscle of a wide variety of species from cold-blooded vertebrates to man. Does not crossreact with smooth or cardiac muscle, non-muscle tissue and cultured cells. Localizes nebulin within the I band of skeletal muscle sections by indirect immunofluorescence.

Immunogen

chicken breast nebulin.

Application

Monoclonal Anti-Nebulin antibody produced in mouse has been used in:
  • immunohistochemistry
  • fluorescent immunostaining
  • western blotting

Biochem/physiol Actions

Nebulin associates with the Z-line and thin actin filament in a sarcomere. It may function as a regulator of sarcomere thin filament length. The protein also acts as a structural link between sarcomeres. Nebulin interacts with the capping protein CapZ and has a role in controlling the localization of actin filaments.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical phenotype, muscle MRI and muscle pathology of LGMD1F
Peterle E, et al.
Journal of Neurology, 260(8), 2033-2041 (2013)
Identification of chicken nebulin isoforms of the 31-residue motifs and non-muscle nebulin.
Joo YM
Biochemical and Biophysical Research Communications (2004)
A 7.5-kb 3'-Terminal cDNA Sequence of Chicken Skeletal Muscle Nebulin Reveals Its Actin Binding Regions.
Suzuki T
Zoological Science (2000)
Nebulin interacts with CapZ and regulates thin filament architecture within the Z-disc.
Pappas CT
Molecular Biology of the Cell (2008)
Biallelic mutations in MYPN, encoding myopalladin, are associated with childhood-onset, slowly progressive nemaline myopathy
Miyatake S, et al.
American Journal of Human Genetics, 100(1), 169-178 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service