Skip to Content
Merck
All Photos(1)

Documents

N8403

Sigma-Aldrich

6-Nitroso-1,2-benzopyrone

ADP-ribosyltransferase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5NO3
CAS Number:
Molecular Weight:
175.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

O=Nc1ccc2OC(=O)C=Cc2c1

InChI

1S/C9H5NO3/c11-9-4-1-6-5-7(10-12)2-3-8(6)13-9/h1-5H

InChI key

HXTDAUGEZTYMGP-UHFFFAOYSA-N

Application

Binds to the DNA-recognizing domain of ADP-ribosyltransferase and preferentially destabilizes one of the two zinc finger polypeptide complexes. The affected enzyme loses almost all activity, but still binds to DNA.
Binds to the DNA-recognizing domain of ADP-ribosyltransferase and preferentially destabilizes one of the two zinc finger polypeptide complexes. The affected enzyme loses almost all activity, but still binds to DNA.>

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy G Owen et al.
The Biochemical journal, 436(3), 709-717 (2011-04-07)
PPTases (phosphopantetheinyl transferases) are of great interest owing to their essential roles in activating fatty acid, polyketide and non-ribosomal peptide synthetase enzymes for both primary and secondary metabolism, as well as an increasing number of biotechnological applications. However, existing techniques
Maria Koulmanda et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(42), 16242-16247 (2008-10-15)
Invasive insulitis is a destructive T cell-dependent autoimmune process directed against insulin-producing beta cells that is central to the pathogenesis of type 1 diabetes mellitus (T1DM) in humans and the clinically relevant nonobese diabetic (NOD) mouse model. Few therapies have
K G Buki et al.
FEBS letters, 290(1-2), 181-185 (1991-09-23)
6-Nitroso-1,2-benzopyrone, an oxidation product of 6-amino-1,2-benzopyrone, binds to the DNA-recognizing domain of the ADP-ribose transferase protein and preferentially destabilizes Zn2+ from one of the two zinc finger polypeptide complexes present in the intact enzyme, as determined by the loss of
W G Rice et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(16), 7703-7707 (1992-08-15)
6-Nitroso-1,2-benzopyrone and 3-nitrosobenzamide, two C-nitroso compounds that inactivate the eukaryotic nuclear protein poly(ADP-ribose) polymerase [NAD+:poly(adenosine diphosphate D-ribose) ADP-D-ribosyltransferase, ADPRT, EC 2.4.2.30] at one zinc-finger site, completely suppressed the proliferation of leukemic and other malignant human cells and subsequently produced cell
W G Rice et al.
Nature, 361(6411), 473-475 (1993-02-04)
Retroviral nucleocapsid and gag-precursor proteins from all known strains of retroviruses contain one or two copies of an invariant sequence, Cys-X2-Cys-X4-His-X4-Cys, that is populated with zinc in mature particles. Modification of cysteine or histidine residues results in defective packaging of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service