Skip to Content
Merck
All Photos(1)

Key Documents

N7160

Sigma-Aldrich

NAD-ADH Reagent Multiple Test Vial

for alcohol determination

Synonym(s):

Ethanol assay reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.51
Pricing and availability is not currently available.

Quality Level

form

powder

storage temp.

−20°C

Application

NAD-ADH Reagent Multiple Test Vial has been used to determine the alcohol content in blood.[1] It has also been used to detect ethanol levels in mice.[2]

Biochem/physiol Actions

Alcohol dehydrogenase (ADH) catalyzes the oxidation of alcohol to acetaldehyde[3] with the simultaneous reduction of nicotinamide adenine dinucleotide (NAD) to NADH.[4] The consequent increase in absorbance at 340 nm is directly proportional to alcohol concentration in the sample.

Other Notes

Vial contains ≥10 μmol of nicotinamide adenine dinucleotide (NAD) and ≥800 units of yeast alchohol dehydrogenase (ADH), buffer salts, and stabilizers.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajay C Donepudi et al.
Hepatology communications, 2(1), 99-112 (2018-02-07)
Alcoholic fatty liver disease (AFLD) is a major risk factor for cirrhosis-associated liver diseases. Studies demonstrate that alcohol increases serum bile acids in humans and rodents. AFLD has been linked to cholestasis, although the physiologic relevance of increased bile acids
Ana M Romero et al.
Neurotoxicity research, 29(1), 69-79 (2015-08-13)
Chronic alcohol consumption may cause neurodevelopmental and neurodegenerative disorders. Alcohol neurotoxicity is associated with the production of acetaldehyde and reactive oxygen species that induce oxidative DNA damage. However, the molecular mechanisms by which ethanol disturbs the DNA damage response (DDR)
Tatsuro Kumada et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(3), 742-756 (2006-01-20)
The brains of fetal alcohol syndrome patients exhibit impaired neuronal migration, but little is known about the mechanisms underlying this abnormality. Here we show that Ca2+ signaling and cyclic nucleotide signaling are the central targets of alcohol action in neuronal
Denys V Volgin
Neuroscience letters, 439(2), 182-186 (2008-06-03)
Prenatal alcohol exposure (AE) is associated with lasting abnormalities of sleep and motor development, but the underlying mechanisms are unknown. We hypothesized that AE alters development of GABAergic signaling in the hypothalamic regions important for the control of sleep and
ADH Cluster Genes, Genome-Wide Association Studies, and Alcohol Dependence
Park BL and Shin HD
Neuropathology of Drug Addictions and Substance Misuse, 520-530 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service