N6756
Nicotinamide hypoxanthine dinucleotide, reduced form, sodium salt
Grade I, ≥95%
Synonym(s):
Deamino DPNH, Deamino NADH, Deaminodiphosphopyridine nucleotide, reduced form
About This Item
Recommended Products
type
Grade I
Assay
≥95%
form
powder
storage temp.
−20°C
SMILES string
NC(=O)C1=CN(C=CC1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5C(=O)N=CNc45)C(O)C2O
InChI
1S/C21H28N6O15P2/c22-17(32)9-2-1-3-26(4-9)20-15(30)13(28)10(40-20)5-38-43(34,35)42-44(36,37)39-6-11-14(29)16(31)21(41-11)27-8-25-12-18(27)23-7-24-19(12)33/h1,3-4,7-8,10-11,13-16,20-21,28-31H,2,5-6H2,(H2,22,32)(H,34,35)(H,36,37)(H,23,24,33)
InChI key
WXWNHSQIXJHVJY-UHFFFAOYSA-N
Related Categories
Application
Other Notes
Preparation Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service