Skip to Content
Merck
All Photos(1)

Key Documents

N6414

Sigma-Aldrich

NN-DNJ

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$226.00
25 MG
$785.00

$226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$226.00
25 MG
$785.00

About This Item

Empirical Formula (Hill Notation):
C15H31NO4
CAS Number:
Molecular Weight:
289.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$226.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

CCCCCCCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

InChI

1S/C15H31NO4/c1-2-3-4-5-6-7-8-9-16-10-13(18)15(20)14(19)12(16)11-17/h12-15,17-20H,2-11H2,1H3/t12-,13+,14-,15-/m1/s1

InChI key

FTSCEGKYKXESFF-LXTVHRRPSA-N

Biochem/physiol Actions

NN-DNJ is a glucosidase inhibitor that acts as a pharmacologic chaperone.
NN-DNJ is a glucosidase inhibitor that acts as a pharmacologic chaperone. NN-DNJ also acts synergistically with proteostasis modulators such as celastrol and MG132 to stabilize mutant proteins in properly folded states. NN-DNJ stabilizes various proteins against misfolding, increasing proper trafficking from the endoplasmic reticulum. There is potential use in studying diseases marked by misfolded proteins and has shown antiviral activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service