N5627
Methyl nonanoate
≥98%
Synonym(s):
Methyl pelargonate, Nonanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)7COOCH3
CAS Number:
Molecular Weight:
172.26
Beilstein:
1754521
EC Number:
MDL number:
UNSPSC Code:
12352211
Recommended Products
Assay
≥98%
refractive index
n20/D 1.421 (lit.)
bp
213-214 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CCCCCCCCC(=O)OC
InChI
1S/C10H20O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-9H2,1-2H3
InChI key
IJXHLVMUNBOGRR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service