N4887
Newborn Calf Serum
USA origin, γ-irradiated, sterile-filtered, suitable for cell culture
Synonym(s):
NBCS, NBS, NCS, calf sera, calf serum, newborn calf sera
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352207
Recommended Products
biological source
bovine (calf)
sterility
γ-irradiated
sterile-filtered
origin
USA origin
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Newborn calf serum (NCS) is collected within 10 days post partum calves. NCS is useful in a wide range of cell culture and organ preservation applications. For example, it is used in cornea preservation and cryopreservation of bovine embryos. NCS is used in cell culture to support the maturation of oocytes; to study the function of tubular epithelial cells; and to develop primary cultures of cells such as visceral adipocytes.
Preparation Note
Collected from calves that are 10 days old or less.
Exposed to 25-35 kGy of irradiation.
Exposed to 25-35 kGy of irradiation.
Analysis Note
Endotoxin tested
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ling Chen et al.
American journal of nephrology, 25(5), 459-465 (2005-08-25)
Oxidative stress has been considered to be a common pathogenetic factor of diabetic nephropathy. But the reason why renal cells are susceptible to oxidative injury in diabetes is not clear. Vitamin C plays a central role in the antioxidant defense
Kyoko Shimizu et al.
Cell biology international, 30(4), 381-388 (2006-03-15)
We have recently developed a primary culture system for visceral adipocytes (VAs) using stomal-vascular cells (SVCs) isolated from the mesenteric fat tissue of male Sprague-Dawley rats of 3-5 weeks of age. Modified Dulbecco's modified Eagle medium (DMEM)/F12 containing 17 microM
R Kishida et al.
Theriogenology, 62(9), 1663-1676 (2004-10-30)
To establish a defined in vitro maturation culture system for porcine oocytes, we examined the effects of adding cysteine (Cys) and epidermal growth factor (EGF) to the maturation medium. Furthermore, to evaluate cytoplasmic maturation, we investigated GSH concentrations and embryo
Davide Camposampiero et al.
Cornea, 22(3), 254-258 (2003-03-27)
To explore the possibility of replacing bovine serum with chicken ovalbumin in cornea preservation. Twenty-one pairs of corneas were cultivated at 31 degrees C in a medium containing either 2% (vol/vol) of newborn calf serum (one cornea) or 2 mg/mL
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service