Skip to Content
Merck
All Photos(1)

Key Documents

N4537

Sigma-Aldrich

Nek7, Active human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

NIMA Related Kinase 7, Never in Mitosis Gene A-Related Kinase 7

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$918.00

$918.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$918.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$918.00


Estimated to ship on26 May 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥85% (SDS-PAGE)

form

buffered aqueous glycerol solution

enzyme activity

≥50 units/mg protein

mol wt

protein apparent mol wt ~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK7(140609)

Biochem/physiol Actions

Nek7 is a member of the NIMA (never in mitosis, gene A) family of serine/threonine kinases. During early embryogenesis, Nek7 is expressed in the site of decidual reaction while later in embryogenesis, it is almost exclusively restricted to the nervous system in the dorsal thalamus.

Unit Definition

One unit will incorporate 1 nanomole of phosphate into beta-casein per minute at ph 7.2 at 30 deg C using a final concentration of 50 μm (32p) ATP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service