Skip to Content
Merck
All Photos(1)

Key Documents

N4163

Sigma-Aldrich

Nemadipine-A

≥98% (HPLC), solid

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(pentafluorophenyl)-3,5-pyridinedicarboxylic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18F5NO4
CAS Number:
Molecular Weight:
419.34
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

solid

solubility

H2O: insoluble <2 mg/mL
DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

CCOC(=O)C1=C(C)NC(C)=C(C1c2c(F)c(F)c(F)c(F)c2F)C(=O)OCC

Biochem/physiol Actions

Nemadipine-A is a L-type calcium channel α1-subunit antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service