Skip to Content
Merck
All Photos(1)

Key Documents

N4159

Sigma-Aldrich

NCS-382 hydrate

solid, ≥98% (HPLC)

Synonym(s):

5,7,8,9-Tetrahydro-5-hydroxy-6H-benzocyclohepten-6-ylidene acetic acid monosodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13O3Na · xH2O
CAS Number:
Molecular Weight:
240.23 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: 22 mg/mL

SMILES string

O.[Na+].OC1C(\CCCc2ccccc12)=C\C([O-])=O

InChI

1S/C13H14O3.Na.H2O/c14-12(15)8-10-6-3-5-9-4-1-2-7-11(9)13(10)16;;/h1-2,4,7-8,13,16H,3,5-6H2,(H,14,15);;1H2/q;+1;/p-1/b10-8+;;

InChI key

FCHWTCFJPQOCCJ-PIHABLKOSA-M

Biochem/physiol Actions

γ-Hydroxybutyrate (GHB) receptor antagonist; anti­convulsant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Maitre et al.
The Journal of pharmacology and experimental therapeutics, 255(2), 657-663 (1990-11-01)
Administration of gamma-hydroxybutyrate (GHB) to animals induces electroencephalographic and behavioral changes that resemble petit-mal seizures. Furthermore, these GHB-induced electroencephalogram-behavioral changes can be blocked by anticonvulsant drugs, which are specific in their action against petit-mal seizures. These effects of GHB on
M Maitre
Progress in neurobiology, 51(3), 337-361 (1997-02-01)
gamma-Hydroxybutyrate is a metabolite of GABA which is synthesized and accumulated by neurons in brain. This substance is present in micromolar quantities in all brain regions investigated as well as in several peripheral organs. Neuronal depolarization releases gamma-hydroxybutyrate into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service