Skip to Content
Merck
All Photos(1)

Key Documents

N2165

Sigma-Aldrich

NKY80

≥98% (HPLC)

Synonym(s):

2-Amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N3O2
CAS Number:
Molecular Weight:
229.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

Nc1ncc2C(=O)CC(Cc2n1)c3ccco3

InChI

1S/C12H11N3O2/c13-12-14-6-8-9(15-12)4-7(5-10(8)16)11-2-1-3-17-11/h1-3,6-7H,4-5H2,(H2,13,14,15)

InChI key

SOJUSNIBPPMLCC-UHFFFAOYSA-N

Biochem/physiol Actions

NKY80 is a selective adenylyl cyclase-V inhibitor with greater than 200-fold selectivity for type V over the type II isoform.
NKY80 is a selective adenylyl cyclase-V inhibitor.

Features and Benefits

This compound is a featured product for ADME Tox and Cyclic Nucleotide research. Discover more featured ADME Tox and Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sébastien Didier et al.
International journal of molecular sciences, 19(12) (2018-11-24)
Long-term memory formation depends on the expression of immediate early genes (IEGs). Their expression, which is induced by synaptic activation, is mainly regulated by the 3',5'-cyclic AMP (cAMP)-dependent protein kinase/cAMP response element binding protein (cAMP-dependent protein kinase (PKA)/ cAMP response
Seyedeh Minoo Sajjadian et al.
Open biology, 10(10), 200197-200197 (2020-10-22)
In insect midgut, prostaglandins (PGs) play a crucial role in defending bacterial and malarial pathogens. However, little is known about the PG signalling pathway in the midgut. A dual oxidase (Se-Duox) with presumed function of catalysing reactive oxygen species (ROS)
Sung-Chul Lim et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 21(4), 397-405 (2017-07-15)
MDL-12330A is a widely used adenylyl cyclase (AC) inhibitor that blocks AC/cAMP signaling. In this study, we demonstrated a novel antitumor activity of this drug in gastric carcinoma (GC) cell lines. In these GC cells, MDL-12330A reduced cell viability and
Sören Brandenburg et al.
Journal of molecular and cellular cardiology, 165, 141-157 (2022-01-17)
Axial tubule junctions with the sarcoplasmic reticulum control the rapid intracellular Ca2+-induced Ca2+ release that initiates atrial contraction. In atrial myocytes we previously identified a constitutively increased ryanodine receptor (RyR2) phosphorylation at junctional Ca2+ release sites, whereas non-junctional RyR2 clusters
Adrian P Mansini et al.
Hepatology (Baltimore, Md.), 69(4), 1582-1598 (2018-10-10)
Cholangiocytes, the epithelial cells lining the biliary tree in the liver, express primary cilia that can detect several kinds of environmental signals and then transmit this information into the cell. We have reported that cilia are significantly reduced in cholangiocarcinoma

Articles

Discover Bioactive Small Molecules for ADME/Tox

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service