Skip to Content
Merck
All Photos(1)

Key Documents

N2040

Sigma-Aldrich

2-NP

≥98% (HPLC)

Synonym(s):

2-(1,8-naphthyridin-2-yl)-Phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O
CAS Number:
Molecular Weight:
222.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

orange

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

Oc1ccccc1-c2ccc3cccnc3n2

InChI

1S/C14H10N2O/c17-13-6-2-1-5-11(13)12-8-7-10-4-3-9-15-14(10)16-12/h1-9,17H

InChI key

AYKMXKNVEUMLFQ-UHFFFAOYSA-N

Biochem/physiol Actions

2-NP is a STAT1 Enhancer

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiangyong Liu et al.
Journal of hazardous materials, 403, 123987-123987 (2020-12-04)
Developing highly efficient and cost-effective catalyst with tuned microstructure holds great promise in the reduction of nitroaromatic compounds under mild reaction conditions. Herein, we report a new Co@NC-MF catalyst with a fascinating hierarchical flower-like architecture in situ assembled from uniform
Xueqing Zhou et al.
BMC chemistry, 14(1), 15-15 (2020-02-27)
In this study, a functionalized multiwalled carbon nanotube (MWCNT)-coated solid-phase microextraction (SPME) fiber was developed for concentrating analytes in aqueous samples. Sodium deoxycholate (NaDC) was used as a dispersing agent for non-covalent modification of MWCNTs. The coating showed porous structure
Alina Amirova et al.
Polymers, 12(3) (2020-04-05)
Water-soluble, partially cross-linked poly-2-isopropyl-2-oxazoline combining the properties of chemical and physical gels was synthesized by a two-step procedure. Thermally induced sol-gel transition in its aqueous solution was studied by rheology, light scattering, and turbidimetry. It was demonstrated that the synthesized
Lien Anh Ha Do et al.
The Journal of infectious diseases, 217(1), 134-146 (2017-10-14)
Most insights into the cascade of immune events after acute respiratory syncytial virus (RSV) infection have been obtained from animal experiments or in vitro models. In this study, we investigated host gene expression profiles in nasopharyngeal (NP) swabs and whole
Sudip Nag et al.
Environmental science and pollution research international, 25(3), 2331-2349 (2017-11-11)
Microbial synthesis of gold nanoparticles (GNPs) has attracted considerable attention in recent times due to their exceptional capability for the bioremediation of industrial wastes and also for the treatment of wastewater. A bacterial strain Staphylococcus warneri, isolated from the estuarine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service