Skip to Content
Merck
All Photos(1)

Key Documents

N2010

Sigma-Aldrich

α-Nicotinamide adenine dinucleotide phosphate sodium salt

≥90%

Synonym(s):

α-NADP, α-TPN, α-Triphosphopyridine nucleotide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28N7O17P3
CAS Number:
Molecular Weight:
743.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥90%

impurities

≤1% β-isomer

storage temp.

−20°C

SMILES string

[Na].NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3OP(O)(O)=O)n4cnc5c(N)ncnc45)C(O)C2O

Looking for similar products? Visit Product Comparison Guide

Other Notes

α analog of β-NADP

Caution

This is not the common NADP required in the usual enzyme systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service