Skip to Content
Merck
All Photos(2)

Key Documents

N1786

Sigma-Aldrich

NSC 95397

≥97% (HPLC)

Synonym(s):

2,3-Bis[(2-hydroxyethyl)thio]-1,4-naphthoquinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00

$177.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$177.00

About This Item

Empirical Formula (Hill Notation):
C14H14O4S2
CAS Number:
Molecular Weight:
310.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$177.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

orange, solid

solubility

DMSO: 16 mg/mL

storage temp.

−20°C

SMILES string

OCCSC1=C(SCCO)C(=O)c2ccccc2C1=O

InChI

1S/C14H14O4S2/c15-5-7-19-13-11(17)9-3-1-2-4-10(9)12(18)14(13)20-8-6-16/h1-4,15-16H,5-8H2

InChI key

MAASHDQFQDDECQ-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Irreversible Cdc25 dual specificity phosphatase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qin Zhou et al.
PloS one, 5(10), e13538-e13538 (2010-11-05)
Keratin polypeptide 19 (K19) is a type I intermediate filament protein that is expressed in stratified and simple-type epithelia. Although K19 is known to be phosphorylated on tyrosine residue(s), conclusive site-specific characterization of these residue(s) and identification potential kinases that
Lei Jin et al.
Communications biology, 3(1), 349-349 (2020-07-06)
Angiogenesis is required for tissue repair; but abnormal angiogenesis or neovascularization (NV) causes diseases in the eye. The avascular status in the cornea is a prerequisite for corneal clarity and thought to be maintained by the equilibrium between proangiogenic and
Hong Li et al.
International journal of biological sciences, 16(7), 1107-1120 (2020-03-17)
Traumatic brain injury (TBI) induces an acute inflammatory response in the central nervous system that involves both resident and peripheral immune cells. The ensuing chronic neuroinflammation causes cell death and tissue damage and may contribute to neurodegeneration. The molecular mechanisms
Yan Han et al.
Journal of proteome research, 17(1), 579-589 (2017-12-21)
The new strategy for chemical toxicity testing and modeling is to use in vitro human cell-based assays in conjunction with quantitative high-throughput screening (qHTS) technology, to identify molecular mechanisms and predict in vivo responses. Stem cells are more physiologically relevant
Xun Chen et al.
Molecules and cells, 43(6), 517-529 (2020-05-22)
Carboxyl-terminal binding proteins (CtBPs) are transcription regulators that control gene expression in multiple cellular processes. Our recent findings indicated that overexpression of CtBP2 caused the repression of multiple bone development and differentiation genes, resulting in atrophic nonunion. Therefore, disrupting the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service