Skip to Content
Merck
All Photos(2)

Key Documents

N1530

Sigma-Aldrich

Nomifensine maleate salt

Synonym(s):

1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$401.00

$401.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$401.00

About This Item

Empirical Formula (Hill Notation):
C16H18N2 · C4H4O4
CAS Number:
Molecular Weight:
354.40
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$401.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CN1CC(c2ccccc2)c3cccc(N)c3C1

InChI

1S/C16H18N2.C4H4O4/c1-18-10-14(12-6-3-2-4-7-12)13-8-5-9-16(17)15(13)11-18;5-3(6)1-2-4(7)8/h2-9,14H,10-11,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

GEOCVSMCLVIOEV-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nomifensine maleate salt has been used to study the β-CFT (cocaine analogue) binding using human embryonic kidney 293T cells.[1]

Biochem/physiol Actions

Nomifensine maleate is a selective dopamine uptake inhibitor interacting with the dopamine transporter,[2] at a site different from that of cocaine; antidepressant. Nomifensine serves as an antidepressant and is known to cause frequent hemolytic anemia. The aniline group in this compound results in blood and liver toxicities.[3]

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of nomifensine to a dihydroisoquinolinium ion metabolite by human myeloperoxidase, hemoglobin, monoamine oxidase-A, and cytochrome P450 enzymes
Obach RS and Dalvie DK
Drug Metabolism and Disposition (2006)
Yoonbae Oh et al.
Biosensors & bioelectronics, 121, 174-182 (2018-09-16)
For over two decades, fast-scan cyclic voltammetry (FSCV) has served as a reliable analytical method for monitoring dopamine release in near real-time in vivo. However, contemporary FSCV techniques have been limited to measure only rapid (on the order of seconds
Matthew Dunn et al.
Nature communications, 9(1), 2838-2838 (2018-07-22)
Norepinephrine is a monoamine neurotransmitter with a wide repertoire of physiological roles in the peripheral and central nervous systems. There are, however, no experimental means to study functional properties of individual noradrenergic synapses in the brain. Development of new approaches
S M Meiergerd et al.
Journal of neurochemistry, 63(5), 1683-1692 (1994-11-01)
The inhibition by cocaine of the apparent initial rate of the transport of striatal dopamine was compared with inhibitions produced by cocaethylene, benztropine, GBR-12909, mazindol, and nomifensine. Rotating disk electrode voltammetry was used to measure the kinetically resolved, inwardly directed
Kioko Guzmán-Ramos et al.
Learning & memory (Cold Spring Harbor, N.Y.), 19(10), 453-460 (2012-09-18)
Previous findings indicate that the acquisition and consolidation of recognition memory involves dopaminergic activity. Although dopamine deregulation has been observed in Alzheimer's disease (AD) patients, the dysfunction of this neurotransmitter has not been investigated in animal models of AD. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service