Skip to Content
Merck
All Photos(1)

Key Documents

N1018

Sigma-Aldrich

3-Nitro-L-tyrosine ethyl ester hydrochloride

≥99% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O5 · HCl
CAS Number:
Molecular Weight:
290.70
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (titration)

form

powder

color

yellow

mp

185 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl.CCOC(=O)[C@@H](N)Cc1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C11H14N2O5.ClH/c1-2-18-11(15)8(12)5-7-3-4-10(14)9(6-7)13(16)17;/h3-4,6,8,14H,2,5,12H2,1H3;1H/t8-;/m0./s1

InChI key

GITKQXFBNJTMRP-QRPNPIFTSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

3-Nitro-L-tyrosine ethyl ester hydrochloride or NTEE is used measure enzyme activity following carboxylate modification.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guinevere Strack et al.
The journal of physical chemistry. B, 113(35), 12154-12159 (2009-08-12)
The biochemical system logically processing biochemical signals using immune-specific and biocatalytic reactions was designed, and the generated output signals were analyzed by AFM and optical means. Different patterns of immune signals resulted in the formation of various interfacial structures followed
S K Niture et al.
European journal of biochemistry, 268(3), 832-840 (2001-02-13)
Fusarium moniliforme NCIM 1276 isolated from a tropical mangrove ecosystem produces a single extracellular endo-polygalacturonase with an M(r) of 38 kDa and a carbohydrate content of 4%. It has an alkaline pI of 8.1. The K(m) is 0.12 mg.mL(-1), V(max)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service