Skip to Content
Merck
All Photos(8)

Key Documents

MX633S20

Sigma-Aldrich

Mix-n-Stain CF 633 Antibody Labeling Kit (5-20μg)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$193.00

$193.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$193.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$193.00


Check Cart for Availability

form

liquid

Quality Level

storage condition

protect from light

fluorescence

λex 630 nm; λem 650 nm

storage temp.

−20°C

General description

This labeling kit is for labeling 5-20 μg antibody with the far red fluorescent dye CF633. Mix-n-Stain CF dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. CF633 is a novel rhodamine-based far-red dye with excellent fluorescence quantum yield and exceptional photostability. The dye can be efficiently excited by the 633 or 635 nm laser with emission maximum at 650 nm. Both the excitation and emission wavelengths of CF633 are shorter than those of the cyanine-based dye Cy®5 or Alexa Fluor 647. Despite the spectral difference, CF633 is still brighter than Alexa Fluor 647 and other similar cyanine dyes when used in flow cytometry applications on popular flow cytometers. However, on other instruments where the detection windows are more optimized for Cy®5-like dyes, the full advantage of CF633 may not be evident. Ensure that instrument settings are optimized for use with CF633 to obtain the most benefit from the dye. The most important advantage of CF633 is its exceptional photostability, which is unmatched by any of the cyaninebased far-red dyes, including Cy®5, Alexa Fluor 647 and DyLight® 649.

Application

The Mix-n-Stain CF dye antibody labeling kit can be used to label primary antibody for ELISA, western blot, immunohistochemistry, flow cytometry, immunofluorescence or immunocytochemistry applications.

Features and Benefits

Mix-n-Stain CF dye antibody labeling kits dramatically simplify the process of preparing fluorescently labeled antibodies, particularly primary antibodies. Simply mix your antibody with a CF dye of your choice in the buffer provided, a step that takes less than 30 seconds of hands-on time. After waiting for another 30 minutes, without a separation step, you will have a fluorescent antibody conjugate that is as good as the best pre-labeled fluorescent antibody you can buy from some of the leading suppliers. There is no need to calculate how much dye you should use; just mix your antibody with the entire amount of dye provided and you will always produce optimal labeling. Moreover, the labeling reaction can tolerate the presence of common stabilizers, such as sodium azide, Tris, glycerol and low level of BSA.

Components

The kit consists of 1 vial each of Mix-n-Stain Dye, Mix-n-Stain Reaction Buffer (10x), Mix-n-Stain Storage Buffer, and an ultrafiltration vial.

Analysis Note

Direct replacement for: Alexa Fluor 633, DY-630, DyLight® 633

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Cy is a registered trademark of Cytiva
DyLight is a registered trademark of Pierce Biotechnology, Inc.
Mix-n-Stain is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service