Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1300

Sigma-Aldrich

Canine MDCKII Wild Type Cells

canine

Synonym(s):

MDCK II, MDCK-2, MDCKII-WT

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,910.00

$1,910.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$1,910.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

$1,910.00


Check Cart for Availability

Request a Bulk Order

Product Name

Canine MDCKII Wild Type Cells,

biological source

canine

Quality Level

form

liquid

storage temp.

−196°C

General description

MDCK II - Madin-Darby canine kidney- is a subclone derived from the heterogenous parent line - MDCK (ECACC catalogue no. 85011435).

Application

Canine MDCKII Wild Type Cells can be used as an experimental model to study the generation and maintenance of cell surface polarity in epithelial cells.
The cell line can be used as an experimental model to study the generation and maintenance of cell surface polarity in epithelial cells. See technical bulletin for detailed protocols.

Features and Benefits

MDCK II - Madin-Darby canine kidney- is a subclone derived from the heterogenous parent line - MDCK (ECACC catalogue no. 85011435) MDCK II cells, which predominate in later passages from MDCK, are reported to display electrical resistance of 100 ohm/cm2. This strain is thought to be derived from the distal tubule or collecting duct of the nephron.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification.

Storage and Stability

Upon receiving a shipment of frozen canine MDCKII wild type cells it is important the end user gives the shipment attention without delay. To ensure the highest level of viability, thaw the vial and initiate the culture as soon as possible upon receipt. If upon arrival, continued storage of the frozen culture is necessary, it should be stored in liquid nitrogen vapor phase and not at –70 °C. Storage at –70 °C will result in loss of viability.

Legal Information

Exhibit 1: ADME/Tox cell lines licenseMDCKII subclone was originally isolated by Daniel Louvard, Institut Curie, Paris France. [1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Louvard
Proceedings of the National Academy of Sciences of the United States of America, 77(7), 4132-4136 (1980-07-01)
A dog kidney epithelial cell line (MDCK), grown in monolayer, displayed in vitro an asymmetric localization of surface proteins. Aminopeptidase [alpha-aminoacylpeptide hydrolase (microsomal), EC 3.4.11.2] was found only in the apical face whereas Na+, K+-ATPase (ATP phosphohydrolase, EC 3.6.1.3) was
G C Hansson et al.
The EMBO journal, 5(3), 483-489 (1986-03-01)
The glycosphingolipids (GSLs) of two sublines of Madin-Darby canine kidney (MDCK) cells, an epithelial cell line, were characterized by t.l.c., antibody overlay and mass spectrometry. The major characteristic which distinguishes the two MDCK cell strains is their trans-epithelial electrical resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service