MTCT9009
MTC™ Bio Strainer Cap for FlowTubes™
pore size 35 μm, green, non-sterile, case of 500 ea (20 bags of 25ea)
Synonym(s):
cell strainer, cell filter cap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
green
low-density polyethylene
sterility
non-sterile
packaging
case of 500 ea (20 bags of 25ea)
manufacturer/tradename
MTC™ Bio T9009
pore size
35 μm
General description
Strainer Cap for FlowTubes™ (cap only), with 35µm strainer mesh, dual position for 12x75mm tubes, sterile, 20 bags of 25 caps, 500/pack
LDPE Bio Strainer cap contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications during collection of dissociated samples for subsequent processing in automated flow cytometry instruments.
LDPE Bio Strainer cap contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications during collection of dissociated samples for subsequent processing in automated flow cytometry instruments.
Features and Benefits
- Compatible with FACS (BD) instruments
- Available with 35µm strainer cap, standard cap, or no cap
- Clear polystyrene
- 5mL capacity
Legal Information
FlowTubes is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service