MSST0040
SILu™Lite IFNG Interferon Gamma human
recombinant, expressed in HEK 293 cells, MS Protein Standard
Synonym(s):
IFNγ, IFN-gamma, Immune interferon, Immuneinterferon
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in HEK 293 cells
Assay
≥98% (SDS-PAGE)
form
lyophilized powder
potency
≥98% Heavy amino acids incorporation efficiency by MS
mol wt
calculated mol wt 17 kDa
technique(s)
mass spectrometry (MS): suitable
suitability
suitable for mass spectrometry (internal calibrator)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... IFNG(3458)
General description
Immunogen
Biochem/physiol Actions
Physical form
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service