Skip to Content
Merck
All Photos(1)

Key Documents

MSST0015

Sigma-Aldrich

SILuProt B2M beta-2-microglobulin human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C- and 15N-labeled

Synonym(s):

β-2-microglobulin

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$1,340.00

$1,340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$1,340.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

$1,340.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... B2M(567)

General description

SILu Prot B2M is a recombinant, stable isotope-labeled human B2M which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of B2M in mass-spectrometry. SILu Prot B2M is a monomer of 119 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~14 kDa.
Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)

Biochem/physiol Actions

B2M is the light chain of the major histocompatibility class (MHC) I molecule expressed on the cell surface of all nucleated cells. Increased urinary B2M excretion has been observed to be an early marker of tubular injury in a number of settings, including nephrotoxicant exposure, cardiac surgery, and renal transplantation, preceding rises in serum creatinine by as many as 4−5 days. B2M may also serve as an early biomarker for AKI (Acute Kidney Injury).

Sequence

IQRTPKIQVYSRHPAENGKSNFLNCYVSGFHPSDIEVDLLKNGERIEKVEHSDLSFSKDWSFYLLYYTEFTPTEKDEYACRVNHVTLSQPKIVKWDRDMDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service