Skip to Content
Merck
All Photos(1)

Key Documents

MSST0006

Sigma-Aldrich

SILuLite VEGFA Vascular endothelial growth factor A human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

SILuLite Vascular endothelial growth factor A

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$1,330.00

$1,330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$1,330.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

$1,330.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... VEGFA(7422)

General description

SILu Lite VEGF165 is a recombinant glycosylated human protein expressed in human 293 cells. It is a homodimer with an apparent molecular mass of 45 kDa.


Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)

Biochem/physiol Actions

VEGF165 belongs to the PDGF/VEGF growth factor family characterized by the presence of eight conserved cysteine residues and a cystine knot structure.1 VEGF is secreted by the majority of tumor cells and initiates angiogenesis by activating endothelial cells of existing blood vessels and promoting their migration.2
VEGF has also been implicated in correlation with poor prognosis in breast cancer.2 In addition, VEGF is released in rheumatoid arthritis in response to TNF-α, increasing endothelial permeability and stimulating angiogenesis (formation of capillaries)3,4

Sequence

APMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGC

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service