Skip to Content
Merck
All Photos(1)

Documents

MSQC12

Sigma-Aldrich

SILuCHOP Stable-Isotope Labeled CHO Proteins

CHO Host Cell Protein Mixture

Synonym(s):

Labeled CHO Proteins, SILu CHO Proteins, Stable-Isotope Labeled Proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.24

Quality Level

packaging

vial of 100 μg

suitability

suitable for
suitable for mass spectrometry

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Contamination of final product with Host Cell Proteins (HCPs) is a major concern for the development and production of Biotherapeutics in CHO cell lines. SILuCHOP is a representative heavy-isotope labeled standard of secreted host cell proteins, thus facilitating identification of biotherapeutics with contaminant host cell proteins by mass spectrometry.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
SILu is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Residual presence of host cell proteins (HCPs) in recombinant therapeutic products has considerable clinical safety risks associated with a potential immunological response in patients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service