Skip to Content
Merck
All Photos(2)

Key Documents

MSP15

Sigma-Aldrich

Membrane Scaffold Protein 1D1 Delta-Helix 5

recombinant, expressed in E. coli, Delta Helix 5 deleted

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$843.00

$843.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$843.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

$843.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥90% (SDS-GE)

form

buffered aqueous solution

shipped in

ambient

storage temp.

−20°C

General description

Nanodisc technology is an approach to render membrane proteins soluble in aqueous solutions in a native-like bilayer environment, where the membrane proteins remain stable and active. The Nanodisc concept is derived from high density lipoprotein (HDL) particles and their primary protein component, apolipoprotein. The Nanodisc is a non-covalent structure of phospholipid bilayer and membrane scaffold protein (MSP), a genetically engineered protein, which mimics the function of Apolipoprotein A-1 (ApoA-1).

The first MSP, MSP1, was engineered with its sequence based on the sequence of A-1, but without the globular N-terminal domain of native A-1. The Membrane Scaffold Protein 1D1 (MSP1D1) variant of MSP1 deletes the first 11 amino acids in the Helix 1 portion (referred to as “H0.5” in the accompanying figure) of the original MSP1 sequence. The MSP1D1ΔH5 variant of MSP1D1 omits the 22 amino acids of the Delta-Helix 5 sequence (H5).

Application

For guidelines on the use of this and other MSP′s to prepare Nanodiscs, please visit our Protocols for Membrane Scaffold Proteins and Nanodisc Formation page.

Legal Information

Nanodisc technology, and many of its uses, are covered by the following patents held by the University of Illinois.
  • 7,691,414 Membrane scaffold proteins
  • 7,662,410 Membrane scaffold proteins and embedded membrane proteins
  • 7,622,437 Tissue factor compositions and methods
  • 7,592,008 Membrane scaffold proteins
  • 7,575,763 Membrane scaffold proteins and tethered membrane proteins
  • 7,083,958 Membrane scaffold proteins
  • 7,048,949 Membrane scaffold proteins

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Nanodisc technology aids membrane protein solubilization, overcoming associated challenges in diverse protein classes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service