Skip to Content
Merck
All Photos(2)

Key Documents

MBD0058

Sigma-Aldrich

Phosphate Buffered Saline

1X solution, liquid, pH 7.5, microbial DNA free

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$33.00
5 X 1 ML
$124.00

$33.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$33.00
5 X 1 ML
$124.00

About This Item

UNSPSC Code:
41141527
NACRES:
NA.25

$33.00


Check Cart for Availability

Request a Bulk Order

Product Name

Phosphate Buffered Saline, DNase free, microbial DNA free, 1X solution, pH 7.5

Quality Level

form

liquid

specific activity

(Microbial DNA free buffer)

packaging

pkg of 5X1 mL
pkg of 1 mL

pH

7.5

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Phosphate-buffered saline (PBS) is a buffer solution used in biological research. It is a water-based salt solution containing sodium phosphate, sodium chloride, potassium chloride and potassium phosphate.

Application

Phosphate buffered saline, DNA and DNase free can be used to resuspend Metapolyzyme, DNA free (MAC4LDF) or other lytic enzyme preparations used before DNA extraction. It is supplied in convenient ready-made, liquid form, in 1 mL tubes suitable for UV irradiation.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service