Skip to Content
Merck
All Photos(1)

Key Documents

MBD0027

Sigma-Aldrich

Genomics standardization kit from Porphyromonas gingivalis

Suitable for PCR, sequencing and NGS

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$789.00

$789.00


Estimated to ship on18 May 2025



Select a Size

Change View
1 KIT
$789.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.24

$789.00


Estimated to ship on18 May 2025


Quality Level

concentration

>10^8

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses.1 Genomics standardization kit from Porphyromonas gingivalis can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs.

P. gingivalis, is a gram-negative anaerobe bacterium which is an etiologic agent of adult periodontitis, a chronic inflammatory disease characterized by the destruction of the supportive tissue surrounding teeth. P. gingivalis usually inhabits the oral cavity, being a member of the oral microbiome along with more than 500 other bacterial species.2 Studies have shown that LPS from P. gingivalis plays an important role in this disease. 3-6 The association of the oral microbiota, including P. gingivalis, with various pathological states has been reported. These include development of Alzheimer′s disease7, role in oral cancers8, preterm birth9 and rheumatoid arthritis.10 Studies show that in mice, oral administration of P. gingivalis induced a change in gut microbiota composition, which in turn affects the host′s metabolic profile. Accumulating evidence suggests a profound effect of altered gut metabolite profiles on overall host health.11

View all Microbiome inactivated bacteria and DNA standard for your Metagenomics Research needs.

Application

Suitable for DNA extraction, PCR, sequencing, next generation sequencing

Features and Benefits

  • Individual microbial standard for microbiomics and meta-genomics workflow
  • Suitable standard for PCR, sequencing and NGS
  • Improve Bioinformatics analyses
  • Increases reproducibility
  • Compare results lab to lab

Components

1. Microbial DNA standard from Porphyromonas gingivalis: The genomic DNA is provided at =10 ng/μl concentration in TE buffer pH 8.0. It is recommended to avoid freeze thaw cycles of this product.
2. Inactivated Porphyromonas gingivalis: The inactivated bacteria are provided at >10^8 bacteria/vial as lyophilized powder. It is recommended to avoid freeze thaw cycles of this product.
3. Resuspension Buffer: The Resuspension Buffer is used to reconstitute the lyophilized inactivated bacteria prior to genomic DNA extraction

Preparation Note

Inactivated Porphyromonas gingivalis

1. Thaw the lyophilized cells at RT for 5 min.
2. Use 1 ml of the supplied Resuspension buffer to resuspend the lyophilized cells, pipette until all the cells are suspended.
3. Transfer the resuspended cells to a clean 1.5ml Eppendorf tube.
4. The resuspended bacteria can be used as control and/or spike-in for gDNA extraction from microbiome samples.

It is recommended to avoid long-term storage of the resuspended cells.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service