The inactivation method is proprietary, and sterility testing is conducted to ensure that complete inactivation has been achieved.
MBD0017
Inactivated Escherichia coli
Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/ml
Select a Size
About This Item
Quality Level
form
liquid
concentration
>10^8 bacteria/ml
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
suitability
suitable for restriction endonuclease digests, PCR amplification, Southern blots, and sequencing reactions
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
Suitable for Quantitative standard for PCR, Sequencing and NGS
Features and Benefits
- Individual microbial standard for microbiomics and meta-genomics workflow
- Suitable standard for PCR, sequencing and NGS
- Improve Bioinformatics analyses
- Increases reproducibility
- Compare results lab to lab
Physical form
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.
-
Can you provide details on how the bacteria were inactivated? Were they chemically denatured or heat inactivated?
1 answer-
Helpful?
-
-
What is the concentration of the bacteria in the suspension?
1 answer-
The number of inactivated bacteria per vial is estimated by the CFU count of the untreated control and not by direct count of the final product. The > 10x8 number indicated is an estimation of the number of inactivated bacterial cells per vial.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service