Skip to Content
Merck
All Photos(1)

Key Documents

MBD0009

Sigma-Aldrich

Inactivated Porphyromonas gingivalis

Suitable for DNA extraction, PCR, sequencing, next generation sequencing, >10^8 bacteria/vial

Synonym(s):

Standard DNA

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$472.00

$472.00


Check Cart for Availability


Select a Size

Change View
1 ML
$472.00

About This Item

UNSPSC Code:
41105500
NACRES:
NA.51

$472.00


Check Cart for Availability

Quality Level

form

powder

concentration

>10^8 bacteria/vial

technique(s)

DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Standardization of sample analysis is currently needed in microbiome genomics research workflow. Lack of standardization can lead to biases and errors in common processes during sample preparation and analysis such as sample amplification, sequencing and bioinformatics analyses. Inactivated Prophyromonas gingivalis can serve as standard for benchmarking the performance along the workflow of microbiomics or meta-genomics analyses and as a tool to increase reproducibility and allow comparison of results obtained by different labs. Porphyromonas gingivalis, is a gram negative bacterium which is an etiologic agent of adult periodontitis, a chronic inflammatory disease characterized by the destruction of the supportive tissue surrounding teeth. Studies have shown that LPS from P. gingivalis plays an important role in this disease. The association of the oral microbiota, including P. gingivalis, with various pathological states has been reported. These include development of Alzheimer’s disease, role in oral cancers, preterm birth and rheumatoid arthritis.

Read here how to use our standards to ensure data integrity for your microbiome research.

Application

Inactivated bacteria are provided at >10^8 bacteria/vial as lyophilized powder. It is recommended to avoid freeze thaw cycles of this product.

Physical form

Powder -The inactivated bacteria are provided at >10^8 bacteria/vial as lyophilized powder

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Turn to our Next-Gen Sequencing Oligos (NGSO) as a potential solution if you are experiencing high levels of adapter dimer formation with custom adapters sourced from other vendors.

DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.

Questions

  1. What does "inactivated Pg" mean?

    1 answer
    1. Inactivated refers to rendering something non-viable or dead. Researchers who purchase products from this category typically seek to work with a complete bacterium (intact DNA) but without dealing with a live bacterium. Certain labs seek to understand how to deactivate the bacterium without harming its DNA, enabling them to obtain the product internally. As a result, the inactivation method is kept confidential. Sterility testing is conducted to confirm that complete inactivation has been accomplished.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service