Skip to Content
Merck
All Photos(1)

Key Documents

MAS007

Sigma-Aldrich

Histamine Quick Test Strips

Sufficient for 10 tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$153.00

$153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$153.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.84

$153.00


Check Cart for Availability

Request a Bulk Order

input

beverage(s)
food(s)

application(s)

food and beverages

detection method

colorimetric

storage temp.

2-8°C

General description

Histamine (C5H9N3) is a biogenic amine naturally present in many foods, fermented products and body cells in amounts without toxicological significance. It is also a contaminant mostly found in Scombridae family fishes, such as tuna and mackerel, as well as other seafood products when they are improperly handled or stored. The consumption of foods containing high levels of histamine may lead to an allergy-like food poisoning known as scombroid poisoning. Therefore, monitoring histamine levels is essential for ensuring food safety and preventing foodborne illnesses, maintaining product quality, and shelf-life.

Application

The Histamine Quick Test Strips are designed for determining histamine levels in the following applications:
  • Food and Beverage Testing
  • Seafood Testing

Features and Benefits

  • Rapid and Sensitive: Delivers swift results with the option of using either 250 μL of sample or 500 mg for fish samples, enabling semi-quantitative measurement across a range of 0-200 ppm
  • Time-Saving: Enables sample treatment and assay completion in under 15 minutes, ensuring efficient testing without compromising accuracy, saving valuable time for users.
  • User-Friendly Design: Simplifies testing with a convenient test strip design, eliminating the need for a plate or cuvette reader, enhancing accessibility and usability for individuals of all skill levels.

Suitability

This kit is suitable for the detection of histamine in food and beverage samples.

Principle

The Histamine Quick Test Strips are based on the histamine dehydrogenase catalyzed oxidation of histamine in which the formed electron mediator reduces a formazan (MTT) reagent. The intensity of the product color is directly proportional to histamine concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service