Skip to Content
Merck
All Photos(1)

Key Documents

MAK532

Sigma-Aldrich

Nitric Oxide Synthase (NOS) Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$792.00

$792.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$792.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$792.00


Check Cart for Availability

Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

cardiovascular diseases; immunological diseases; aging/geriatric diseases; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Nitric oxide (NO) is a reactive radical that plays an important role in many key physiological functions. NO, an oxidation product of arginine by nitric oxide synthase (NOS), is involved in host defense and development, activation of regulatory proteins and direct covalent interaction with functional biomolecules.

Features and Benefits

  • Linear detection range: 0.25 - 25 U/L

Suitability

The kit is suitable for the determination of nitric oxide synthase activity in biological samples, as well as for studying the effects of drugs on nitric oxide synthase activity.

Principle

The Nitric Oxide Synthase (NOS) Assay Kit nvolves two steps: a NOS reaction step during which NO is produced followed by an NO detection step. Since the NO generated by NOS is rapidly oxidized to nitrite and nitrate, the NO production is measured following reduction of nitrate to nitrite using an improved Griess method. The procedure is simple and the time required for sample pretreatment and assay is reduced to as short as 40 minutes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service