Skip to Content
Merck
All Photos(1)

Key Documents

MAK517

Sigma-Aldrich

Glutathione (GSH) Assay Kit

sufficient for 250 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$728.00

$728.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$728.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$728.00


Check Cart for Availability

Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

cancer

shipped in

ambient

storage temp.

2-8°C

General description

Glutathione is a tripeptide of glycine, glutamic acid and cysteine. In the red blood cell, the reduced form of glutathione is vital in maintaining hemoglobin in a reduced state and hence protecting the cells from oxidative damage. Glutathione is involved in detoxification of hydrogen peroxide through glutathione oxidase. Low levels of glutathione are found in deficiencies of key enzymes involved in glutathione metabolism, such as glucose-6-phosphate dehydrogenase, glutathione synthase and glutathione reductase.

Features and Benefits

  • Linear detection range: 0.4 - 100 µM
  • No interference from amino acids and common buffers.

Suitability

The kit is suitable for the determination of reduced glutathione in whole blood, plasma, serum, urine, tissue, and cell extracts, as well as for studying the effects of drugs on glutathione metabolism.

Principle

The Glutathione (GSH) Assay Kit is designed to accurately measure reduced glutathione in biological samples. The improved 5,5′-dithiobis(2-nitrobenzoic acid (DTNB) method combines deproteination and detection into one reagent. DTNB reacts with reduced glutathione to form a yellow product. The optical density, measured at 412 nm, is directly proportional to glutathione concentration in the sample. The optimized formulation has a long shelf life and is completely free of interference due to sample turbidity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service