Skip to Content
Merck
All Photos(1)

Key Documents

MAK512

Sigma-Aldrich

Malate Dehydrogenase Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$817.00

$817.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$817.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.84

$817.00


Check Cart for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

relevant disease(s)

Alzheimer′s disease; neurological disorders; neurodegenerative disorders

shipped in

wet ice

storage temp.

−20°C

General description

Malate dehydrogenase (MDH) is an enzyme which reversibly catalyzes the oxidation of L-malate to oxaloacetate in the presence of NAD. There are 2 isoforms in eukaryotic cells: MDH1 and MDH2. MDH1 is found in the cytoplasm and plays a key part in the malateaspartate shuttle for transporting malate into the mitochondria. MDH2 is a mitochondrial enzyme which participates in the TCA cycle that reversibly converts L-malate into oxaloacetate. Higher MDH activities are found in some neurodegenerative diseases such as Alzheimer′s disease.

Features and Benefits

  • Linear detection range: 0.5 to 65 U/L
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for malate dehydrogenase activity determination in biological samples (e.g. plasma, serum, erythrocytes, tissue and culture media.)

Principle

The Malate Dehydrogenase Assay Kit is a non-radioactive, colorimetric MDH assay based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service