Skip to Content
Merck
All Photos(1)

Key Documents

MAK479

Sigma-Aldrich

NADP/NADPH Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$856.00

$856.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$856.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$856.00


Check Cart for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

General description

Pyridine nucleotides play an important role in metabolism and, thus, there is continual interest in monitoring their concentration levels. Quantitative determination of NADP+ /NADPH has applications in research pertaining to energy transformation and redox state of cells or tissue.

Features and Benefits

  • The linear detection range of the kit is 0.1 µM to 10 µM NADP+ /NADPH
  • Compatible with high-throughput handling systems.
  • Convenient, the procedure involves adding a single working reagent, and reading the optical density at time zero and 30 minutes at room temperature.

Suitability

The kit is suitable for detection of NADP+ /NADPH concentrations and ratios in cell or tissue extracts.

Principle

The NADP/NADPH Assay Kit is based on a glucose dehydrogenase cycling reaction, in which the formed NADPH reduces a formazan (MTT) reagent. The intensity of the reduced product color, measured at 565 nm, is proportionate to the NADP+ /NADPH concentration in the sample. This assay is highly specific for NADP+ /NADPH and is not interfered by NAD+ /NADH.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service