Skip to Content
Merck
All Photos(1)

Key Documents

MAK450

Sigma-Aldrich

Glucose Dehydrogenase Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

GDH Activity Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$760.00

$760.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$760.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$760.00


Check Cart for Availability

Request a Bulk Order

Quality Level

storage condition

(No metal containers. Keep container tightly closed in a dry and well -ventilated place. Keep away from heat and sources of ignition.)

input

tissue
cell culture medium
tissue
plasma

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; hematological disorder

storage temp.

−20°C

General description

Glucose Dehydrogenase (GDH) belongs to the family of oxioreductases, specifically those acting on the CH-OH group of donor with other acceptors. GDH participates in the pentose phosphate pathway.

Application

The Glucose Dehydrogenase Assay Kit may be used for:
  • Metabolism Research
  • Drug Discovery

Features and Benefits

  • Efficient and Precise: Requires only 20 μL of sample and a short 15-minute reaction time while detecting glucose dehydrogenase levels across a broad range from 0.5 to 200 U/L, providing both convenience and sensitivity.
  • Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of D-glucose dehydrogenase enzyme activity and drug effects on its metabolism in biological samples (e.g. plasma, serum, tissue and culture media).

Principle

The Glucose Dehydrogenase Activity Assay Kit is a non-radioactive, colorimetric GDH assay. It is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at A565 nm. The increase in absorbance at A565 nm is proportional to the enzyme activity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service