Skip to Content
Merck
All Photos(1)

Key Documents

MAK436

Sigma-Aldrich

AF Cholesterol Assay Kit

Sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Cholesterol Quantitation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$868.00

$868.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$868.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$868.00


Check Cart for Availability

Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

cardiovascular diseases, depression; cancer; neurological disorders

storage temp.

−20°C

General description

Cholesterol is a sterol and lipid present in the cell membranes, and is transported in the bloodstream of all animals. It is used to form cell membranes and hormones, and plays important roles in cell signaling processes. Elevated levels (hypercholesterolemia) have been associated with cardiovascular diseases such as atherosclerosis; whereas, low levels (hypocholesterolemia) may be linked to depression, cancer and cerebral hemorrhage.

Application

The AF Cholesterol Assay Kit may be used for:
  • Cancer Research
  • Cardiovascular Research
  • Neurological Research

Features and Benefits

Efficient and Precise: Detect Cholesterol levels ranging from 0.1 to 10 mg/dL (colorimetric) and 0.02 to 2 mg/dL (fluorimetric) in a 30 minute room temperature reaction, providing both convenience and sensitivity.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of cholesterol and evaluation of drug effects on cholesterol metabolism in serum, plasma etc.

Principle

Simple, direct and automation-ready procedures for measuring cholesterol are very desirable. The AF Cholesterol Assay Kit uses a single working reagent that combines cholesterol ester hydrolysis, oxidation and color reaction in one step. The color intensity of the reaction product at A570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the total cholesterol concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service