Skip to Content
Merck
All Photos(1)

Key Documents

MAK378

Sigma-Aldrich

Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit

100 fluorometric tests

Synonym(s):

ACE2 Inhibitor Discovery Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$954.00

$954.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$954.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$954.00


Check Cart for Availability

Request a Bulk Order

usage

100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

immunological diseases; cardiovascular diseases; genitourinary disorders; pulmonary disorders

storage temp.

−20°C

General description

Angiotensin II converting enzyme (ACE2), a carboxypeptidase, is part of the renin-angiotensin system (RAS) that controls regulation of blood pressure by cleaving the C-terminal dipeptide of Angiotensin II to convert it into Angiotensin 1-7. It also cleaves Angiotensin I to produce Ang 1-9, of unknown function. ACE2 is a receptor of human coronaviruses, such as SARS and HCoV-NL63. It is expressed on the vascular endothelial cells of heart and kidney. The inhibitors of ACE2 could be able to regulate hypertension by changing vascular permeability. Screening for small molecule and peptide inhibitors might also help in finding treatment for coronavirus mediated infection.

Features and Benefits

The Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit is compatible with high-throughput handling systems.

Suitability

The kit is suitable for Screening/characterizing inhibitors/ligands of ACE2.

Principle

The Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit can be used to screen for potent inhibitors of ACE2 activity, it utilizes the ability of an active ACE2 to cleave a synthetic MCA based peptide substrate to release a free fluorophore. The released MCA can be easily quantified using a fluorescence microplate reader. In the presence of an ACE2 specific inhibitor, the enzyme loses its peptidase activity which results in decrease of fluorescence intensity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 1. For fluorescence measurements, it is supposed to use White flat-bottom 96-well plates or Black flat-bottom 96-well plates? 2. What are the considerations for selecting two time points (T1 & T2) in the linear range of the plot?

    1 answer
    1. This kit requires the use of white flat-bottom 96-well plates. T1 and T2 values are solely dependent on the assay and plate set up. The T1 and T2 should be chosen within the linear range of the kinetic plot of the standard or samples. See the link below for a reference that may be useful.
      https://pubmed.ncbi.nlm.nih.gov/36009234/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service