Skip to Content
Merck
All Photos(1)

Key Documents

MAK225

Sigma-Aldrich

Fatty Acid Methyl Ester Purification Kit

Manufactured by Nacalai Tesque, Inc.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$611.00

$611.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$611.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$611.00


Check Cart for Availability

Request a Bulk Order

usage

sufficient for 50 tests

application(s)

cosmetics
food and beverages
sample preservation

storage temp.

room temp

General description

Fatty Acid Methyl Ester Purification Kit is used to further purify samples prepared with the Fatty Acid Methylation Kit (MAK224). Fatty acid samples are required to be cleaned with this kit before the analysis by a capillary column.

Suitability

Suitable for the purification of methylated fatty acid samples such as blood, yeast, liver, cooking oil, soybean powder, fish oils, etc.

Principle

The Fatty Acid Methylation Kit (Catalog Number MAK224) followed by the Fatty Acid Methyl Ester Purification Kit employs a proprietary reaction technique. Using these kits, the fatty acid methyl esterification/purification process is greatly simplified.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service