Skip to Content
Merck
All Photos(2)

Key Documents

MAK065

Sigma-Aldrich

Lactate Assay Kit II

sufficient for 100 colorimetric tests

Synonym(s):

Lactic Acid Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; cancer

storage temp.

−20°C

General description

L(+)-Lactate is a metabolic compound formed in animals by the action of the enzyme Lactate Dehydrogenase. Lactate is produced in proliferating cells and during anaerobic conditions such as exercise. Abnormally high concentrations of lactate have been related to pathological conditions such as cancer, diabetes, and lactate acidosis. L(+)-Lactate is the major stereoisomer of lactate formed in human intermediary metabolism and is present in blood at levels of around 1-2 mmol/L. This kit provides a convenient means for detecting L(+)-Lactate in biological samples.

Application

Lactate Assay Kit II has been used to measure the levels of lactate in samples.

Suitability

Suitable for detecting L(+)-Lactate in culture medium, fermentation medium, blood and cells.

Principle

Lactate concentration is determined by an enzyme assay, which results in a colorimetric (450 nm) product, proportional t the lactate present. There is no need of pretreatment or purification of samples. Typical sensitivities of detection are between 0.02-10 mM of lactate.

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Jouroukhin et al.
Translational psychiatry, 8(1), 76-76 (2018-04-13)
Our knowledge of how genetic risk variants contribute to psychiatric disease is mainly limited to neurons. However, the mechanisms whereby the same genetic risk factors could affect the physiology of glial cells remain poorly understood. We studied the role of
DISC1 regulates lactate metabolism in astrocytes: implications for psychiatric disorders.
Jouroukhin Y, et al.
Translational Psychiatry, 8(1), 76-76 (2018)
Camaron R Hole et al.
Nature communications, 10(1), 2955-2955 (2019-07-06)
Dendritic cells (DCs), a vital component of the innate immune system, are considered to lack antigen specificity and be devoid of immunological memory. Strategies that can induce memory-like responses from innate cells can be utilized to elicit protective immunity in
Metabolic engineering of Methanosarcina acetivorans for lactate production from methane
McAnulty M J, et al.
Biotechnology and Bioengineering, 114(4), 852-861 (2017)
Nicole J Van Bergen et al.
PloS one, 10(10), e0140919-e0140919 (2015-10-27)
Primary Open Angle Glaucoma (POAG) is a common neurodegenerative disease characterized by the selective and gradual loss of retinal ganglion cells (RGCs). Aging and increased intraocular pressure (IOP) are glaucoma risk factors; nevertheless patients deteriorate at all levels of IOP

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service