Skip to Content
Merck
All Photos(3)

Documents

MAK025

Sigma-Aldrich

Iron Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

cancer; hematological disorder

storage temp.

−20°C

General description

Iron is a mineral that plays an essential role in many biological processes, including DNA synthesis, electron transport and oxygen transport. Iron is a transition element that can form a range of oxidation states, the most common being iron II (Fe2+ or ferrous iron) and iron III (Fe3+ or ferric iron). Iron-containing proteins participate in many reactions, often utilizing transitory changes in the oxidation state of iron to carry out chemical reactions.

Application

Iron assay kit has been used to determine the concentration of Fe2+ in wild type (WT) and Δ ferric uptake regulator (Fur) strains of A. baumannii.

Suitability

Suitable for the measurement of iron concentration in various samples

Principle

In this assay, iron is released by the addition of an acidic buffer. Samples may be tested directly to measure Fe2+ or reduced to measure total iron (Fe2+ and Fe3+). Released iron is reacted with a chromagen resulting in a colorimetric (593 nm) product, proportional to the iron present. The Iron Assay Kit provides a simple convenient means of measuring iron in a variety of biological samples.

Preparation Note

Kit is compatible with serum and tissue samples but not plasma samples.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manman Xu et al.
Journal of cellular physiology, 234(4), 4232-4242 (2018-08-23)
Previous studies have demonstrated an effect of estrogen on iron metabolism in peripheral tissues. The role of estrogen on brain iron metabolism is currently unknown. In this study, we investigated the effect and mechanism of estrogen on iron transport proteins.
Contributions of ferric uptake regulator Fur to the sensitivity and oxidative response of Acinetobacter baumannii to antibiotics
Ajiboye T o, et al.
Microbial Pathogenesis, 119, 35-41 (2018)
Ems T, Huecker MR
Biochemistry, Iron Absorption (2018)
Tippapha Pisithkul et al.
mBio, 10(3) (2019-05-23)
Biofilms are structured communities of tightly associated cells that constitute the predominant state of bacterial growth in natural and human-made environments. Although the core genetic circuitry that controls biofilm formation in model bacteria such as Bacillus subtilis has been well
Jing Ye et al.
Theranostics, 10(26), 12072-12089 (2020-11-19)
Background: Among head and neck squamous cell carcinomas (HNSCCs), hypopharyngeal squamous cell carcinoma (HPSCC) has the worst prognosis. Iron metabolism, which plays a crucial role in tumor progression, is mainly regulated by alterations to genes and post-transcriptional processes. The recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service