Skip to Content
Merck
All Photos(2)

Documents

MAK002

Sigma-Aldrich

L-Amino Acid Quantitation Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

L-Amino Acid Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

storage temp.

−20°C

General description

The new L-Amino Acid Assay Kit, MAK490, is now available!

L-amino acids are essential molecules defined by the nature of their side-chain. There are 20 amino acids that are commonly used by most organisms for the production of proteins and other biologically relevant molecules. While amino acids can occur in either the D- or L-enantimer, only the L-form is used by cells.

Suitability

Suitable for quantitating L-amino acids in body fluids or purified samples.

Principle

In this assay, L-Amino Acid concentration is determined by a coupled enzyme reaction, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the L-Amino Acids present. Glycine is not detectable by this assay.

replaced by

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of endogenous D-amino acid-containing peptides in metazoa.
Bai L, et al.
Bioanalytical reviews., 1(1), 7-24 (2009)
Jinrang Kim et al.
Endocrinology, 160(5), 979-988 (2019-04-03)
Plasma amino acids and their transporters constitute an important part of the feedback loop between the liver and pancreatic α-cell function, and glucagon regulates hepatic amino acid turnover. Disruption of hepatic glucagon receptor action activates the loop and results in
Peng Shang et al.
Aging cell, 16(2), 349-359 (2017-01-14)
The dry (nonneovascular) form of age-related macular degeneration (AMD), a leading cause of blindness in the elderly, has few, if any, treatment options at present. It is characterized by early accumulation of cellular waste products in the retinal pigmented epithelium
Alberts B, et al.
Molecular biology of the Cell. 4th Edition. (2002)
Alberts B, et al.
Molecular Biology in Plant Pathogenesis and Disease Management (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service