Skip to Content
Merck
All Photos(1)

Key Documents

M9555

Sigma-Aldrich

Anti-Matrix Metalloproteinase-9 antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen (active form) 88 kDa
antigen (pro-form) 92 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP9(4318)
mouse ... Mmp9(17395)

Specificity

Reacts with reduced and native MMP-9. The antibody recognizes pro-form and active forms of MMP-9. Mouse MMP-9 is slightly larger than human MMP-9.

Immunogen

corresponding to intact mouse matrix metalloproteinase-9 (gelatinase-B).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Immunohistochemistry-paraffin (1 paper)
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

comparable product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Jönsson et al.
PloS one, 9(8), e105572-e105572 (2014-08-26)
Matrix metalloproteinase (MMP)-9 may play a central role in the development and progression of atherosclerosis. Emerging evidence also indicates an association between MMP-9 and depressive symptomatology. Here, we investigated whether expression of MMP-9 and its inhibitors in blood mononuclear cells
Pei-Chan Zheng et al.
Cancer science, 105(6), 630-638 (2014-04-08)
Calpain small subunit 1 (Capn4) plays a key role in tumor migration or invasion. In this study, expression and function of Capn4 was investigated in human nasopharyngeal carcinoma (NPC). Here we report that both mRNA and protein levels of Capn4
Guo-Cai Li et al.
Cancer science, 101(12), 2546-2553 (2010-10-15)
The effects of mesenchymal stem cells (MSC) on the growth and metastasis of human malignancies including hepatocellular carcinoma (HCC) are controversial, and the underlying mechanisms are not yet understood. The aim of this study was to explore the role of
Spencer Slavin et al.
Carcinogenesis, 35(6), 1301-1309 (2014-01-01)
The prostate cancer (PCa) microenvironment contains active stromal cells known as cancer-associated fibroblasts (CAF) that may play important roles in influencing tumor progression. Here we studied the role of CAF estrogen receptor alpha (ERα) and found that it could protect
Heiani Touaitahuata et al.
Developmental biology, 393(1), 57-70 (2014-07-06)
During long bone development and post-natal growth, the cartilaginous model of the skeleton is progressively replaced by bone, a process known as endochondral ossification. In the primary spongiosa, osteoclasts degrade the mineralized cartilage produced by hypertrophic chondrocytes to generate cartilage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service