M7195
Anti-MO25 (C-terminus) antibody produced in rabbit
~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-CAB39, Anti-CGI-66, Anti-Calcium binding protein 39
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~40 kDa
species reactivity
human, mouse (predicted), rat (predicted)
packaging
antibody small pack of 25 μL
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
concentration
~2 mg/mL
technique(s)
western blot: 0.5-1 μg/mL using HEK-293T cells expressing human MO25 and PC12 cell lysates
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CAB39(51719)
mouse ... Cab39(12283)
rat ... Cab39(301574)
General description
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service